Learn More
Fisherbrand™ Catalyst Norprene Food Formulation Stopped Tubing Assembly
Codice prodotto. p-6877030 Sfoglia Tutto Fisherbrand Traceable Prodotti
Cambia vista
Click to view available options
Diameter (Metric) Inner:
0.8 mm
1.6 mm
3.2 mm
4.8 mm
6.4 mm
8 mm
9.5 mm
Thickness (Metric):
1.6 mm
2.4 mm
Dimensione della confezione:
Pack of 12
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Diameter (Metric) Inner Thickness (Metric) Dimensione della confezione
15511335 0.8 mm 1.6 mm Pack of 12
15521335 1.6 mm 1.6 mm Pack of 12
15531335 3.2 mm 1.6 mm Pack of 12
15541335 4.8 mm 1.6 mm Pack of 12
15551335 6.4 mm 1.6 mm Pack of 12
15561335 8 mm 1.6 mm Pack of 12
15571335 4.8 mm 2.4 mm Pack of 12
15581335 6.4 mm 2.4 mm Pack of 12
15591335 8 mm 2.4 mm Pack of 12
15501345 9.5 mm 2.4 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15511335
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Preconfigured ready-to-use links and assemblies.
  • Available in Platinum-Cured Silicone, TPEs, and PVC
  • Ensures optimal amount of tubing is in pump head
Specifications
Description Stopped tubing assembly
Diameter (Metric) Inner 0.8 mm
Thickness (Metric) 1.6 mm