missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 19 publications
2020.00 NOK - 4200.00 NOK
Specifications
| Antigen | Caspase-9 |
|---|---|
| Dilution | Western Blot reported in scientific literature (PMID 24184144), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 22223358), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen Assay Dependent, Flow (Intracellular) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18758493
|
Novus Biologicals
NB100-56118SS |
0.025 mL |
2020.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18750274
|
Novus Biologicals
NB100-56118 |
0.05 mL |
4200.00 NOK
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspase-9 Polyclonal specifically detects Caspase-9 in Human, Mouse, Rat, Canine, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).Specifications
| Caspase-9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Apoptosis, Cancer, Caspases, Cell Biology, Neurodegeneration | |
| Whole antisera with 0.05% Sodium Azide | |
| APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease | |
| CASP9 | |
| IgG | |
| Unpurified | |
| This antibody detects both the pro- and active-Caspase 9 forms. In western blots, the proform of caspase-9 is detected at ∼50 kDa, the large subunit at ∼35 kDa, and the small subunit at ∼15 kDa. Intermediate caspase-9 cleavage forms may also be seen at ∼21 kDa. |
| Western Blot reported in scientific literature (PMID 24184144), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 22223358), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen Assay Dependent, Flow (Intracellular) | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat, Canine, Gerbil | |
| P55211 | |
| 842 | |
| Human Caspase 9 recombinant protein catalytic subunits (NP_001220). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title