missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 19 publications
Brand: Novus Biologicals NB100-56118
3366.00 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Caspase-9 Polyclonal specifically detects Caspase-9 in Human, Mouse, Rat, Canine, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
Specifications
| Caspase-9 | |
| Polyclonal | |
| Western Blot reported in scientific literature (PMID 24184144), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 22223358), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen Assay Dependent, Flow (Intracellular) | |
| P55211 | |
| CASP9 | |
| Human Caspase 9 recombinant protein catalytic subunits (NP_001220). | |
| 0.05 mL | |
| Alzheimers Research, Apoptosis, Cancer, Caspases, Cell Biology, Neurodegeneration | |
| 842 | |
| Human, Mouse, Rat, Canine, Gerbil | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| This antibody detects both the pro- and active-Caspase 9 forms. In western blots, the proform of caspase-9 is detected at ∼50 kDa, the large subunit at ∼35 kDa, and the small subunit at ∼15 kDa. Intermediate caspase-9 cleavage forms may also be seen at ∼21 kDa. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction