Description
Required when using the cartridge format of Thermo Scientific™ Dionex™ SolEx™ HRP columns. The Dionex SolEx Cartridge Holder V-3 is designed to make it easy to change cartridges when necessary. Dionex SolEx HRP columns and cartridges provide fast, easy sample pre-concentration of contaminants from water samples by solid-phase extraction prior to HPLC analysis. Used online, they are placed in the valve to automatically concentrate the analytes of interest.
Specifications
Specifications
| Description | V-3 Holder |
| For Use With (Equipment) | On-Line SPE Concentration Columns |
| Type | V-3 Cartridge Folder |
| Product Line | SolEx |