Learn More
Hettich™ Carrier for Rotors no. 1624, 1726 and 4758
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 1345
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10426231
  • 840.00 NOK / Each
Estimated Shipment: 25-06-2024
Description
  • Rotor Carrier for Hettich Rotor
  • For use with Rotina, Rotofix and Universal Centrifuges
  • Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
  • ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
  • ROTINA Centrifuges are ideally equipped to deliver separation technology to satisfy the highest standards
Specifications
Specifications
Rotor Adapter
45 mL
100 mm
1
Round Bottom
31 mm
Carriers no. 1624, 1726 and 4758