Learn More
Hettich™ Carrier for Rotors no. 1624, 1726 and 4758
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
840.00 NOK - 980.00 NOK
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
10426231
|
Hettich™
1345 |
45 mL |
840.00 NOK
Each |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12083864
|
Hettich™
1346 |
2 x 20 mL |
840.00 NOK
Each |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10520703
|
Hettich™
1741 |
10 x 1.1 to 9 mL |
980.00 NOK
Each |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Rotor Carrier for Hettich Rotor
- For use with Rotina, Rotofix and Universal Centrifuges
- Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
- ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
- ROTINA Centrifuges are ideally equipped to deliver separation technology to satisfy the highest standards