Learn More
Description
CaptureSelect CH1-XL Affinity Matrix is the improved version of CaptureSelect IgG-CH1 Affinity Matrix. The specificity is the same, but it features a wider elution range (pH 3.0-4.5), increased binding capacity, and improved stability at higher pH.
CaptureSelect CH1-XL Affinity Matrix recognizes the CH1 domain of human IgG antibodies, enabling purification of all subclasses (1, 2, 3, and 4) of human IgG and Fab fragments, independent of the light-chain isotype and source material.
CaptureSelect CH1-XL Affinity Matrix features include:
- Platform approach for purification of IgGs and their Fab fragments directly from complex source materials in a single step with high purity and yield
- Elution possible at pH 4.0–4.5 with 50 mM acetate buffer
- No co-purification of overproduced free light chain due to selectivity for the CH1 domain
- No binding to bovine antibodies, so presence of fetal bovine serum is not a problem
- Excellent scalability
- Non-animal derived
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics:
- Matrix: agarose-based, epoxide-activated
- Average particle size: 65 ± 10 μm
- Ligand: CaptureSelect CH1-XL affinity ligand
- Ligand coupling method: epoxide coupling
- Binding capacity: >15 mg/mL human Fab fragment depending on flow rate, column height, contact time. For humanized Fabs lower capacities can be observed.
- Elution conditions: 100 mM glycine buffer, pH 3.0; 50 mM acetate buffer, pH 4.0–4.5
- Flow rate: 50–200 cm/h (up to 2 bar)
- Formulation buffer: 20% (v/v) ethanol
Specifications
Specifications
| Content And Storage | Store at 2°C to 8°C. |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
| Stationary Phase | Anti-IgG-CH1 |
| Product Line | CaptureSelect |
| Quantity | 50 mL |
| Form | Agarose Bead |
| Particle Size | 65 μm |
| Binding Capacity | >15 mg/mL |
| Flow Rate | 50 to 200 cm/hr. (Up to 2 bar) |
| Ligand Type | CH1-XL affinity ligand |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.