Learn More
Capping Reagent B for DNA Synthesis, Sigma-Aldrich™
Product Code. 32237543
Change view
Click to view available options
Quantity:
450 mL
Unit Size:
450mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32237543 450 mL 450mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32237543 Supplier Sigma Aldrich Supplier No. 8570120450
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
1-Methylimidazole : Tetrahydrofuran 18/82 (w/w), Novabiochem™
Pre-mixed ready-to-use Capping Reagent: 857012.0450: Compatible with MerMade DNA Synthesisers. Attaches directly to Instrument (28/405 neck finish); 857012.4000: Compatible with MerMade DNA Synthesisers. Stand alone from Instrument (38 mm Neck Finish)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Capping Reagent B for DNA Synthesis
Density 0.99 g/cm³
Quantity 450 mL