missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CAMK1G (CaMKI gamma), GST Tag Recombinant Protein
Human CAMK1G (CaMKI gamma), GST Tag Recombinant Protein
Brand: Invitrogen™ A32859
Additional Details : Weight : 0.00900kg
Description
CAMK1G serine/threonine kinase is a member of CAMK family and is a novel membrane-anchored neuronal Ca2+ calmodulin-dependent protein kinase. Full activation of CAMK1G requires both Ca2+ and calmodulin and phosphorylation by CAMKK. CAMK1G transcripts are most abundant in neurons, with the highest levels in limited nuclei such as the central nucleus of the amygdala (CeA) and the ventromedial hypothalamus.- Validated for use with the Z’-LYTE™ assay
- Available in multiple pack sizes for convenient use
Specifications
NP_065172 | |
79.8 kDa | |
CAMK1 Kinase Family | |
Human | |
GST Tag | |
Dry Ice | |
Full Length, Recombinant, Active |
Kinase Assay | |
100 μg | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Insect | |
Liquid |