missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CAMK1G (CaMKI gamma), GST Tag Recombinant Protein

Product Code. p-7180709
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
Product Code. Quantity unitSize
15950273 10 μg 10µg
17158725 100 μg 100µg
17178765 1 mg 1mg
3 options
This item is not returnable. View return policy

Product Code. 15950273

Brand: Invitrogen™ A30489

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This CAMK1G (CaMKI gamma) recombinant human protein (full length) was expressed in insect cells. CAMK1G (CaMKI gamma) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for CAMK1G (CaMKI gamma) is NP_065172.1.

CAMK1G serine/threonine kinase is a member of CAMK family and is a novel membrane-anchored neuronal Ca2+ calmodulin-dependent protein kinase. Full activation of CAMK1G requires both Ca2+ and calmodulin and phosphorylation by CAMKK. CAMK1G transcripts are most abundant in neurons, with the highest levels in limited nuclei such as the central nucleus of the amygdala (CeA) and the ventromedial hypothalamus.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96NX5
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 10mM glutathione, 0.1mM EDTA, 25% glycerol, 150mM NaCl, 0.1mM PMSF, 0.25mM DTT and no preservative; pH 7.5
Gene ID (Entrez) 57172
Molecular Weight (g/mol) 79.8 kDa
Name Human CAMK1G (CaMKI gamma), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias calcium/calmodulin dependent protein kinase IG; calcium/calmodulin-dependent protein kinase I gamma; calcium/calmodulin-dependent protein kinase IG; calcium/calmodulin-dependent protein kinase type 1G; CaM kinase 1G; CaM kinase I gamma; caM kinase IG; CAMK1G; caMKI gamma; caM-KI gamma; caMKIG; CaMKIgamma; caMKI-gamma; caMK-like CREB kinase III; CLICK III; CLICK3; CLICKIII; CLICK-III; dJ272L16.1; RP1-272L16.2; VWS1
Common Name CaMKI gamma
Gene Symbol Camk1g
KinaseFamily CAMK1 Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.