Learn More
Hanna Instruments™ Calibration Check Key
Designed to check the accuracy of thermistor thermometers that use HI 765 series probes. The Hanna Instruments™ Calibration Check Key offers a fast and simple way of checking the accuracy of your instrument to within ±0.4°C.
Marca: Hanna Instruments™ HI765000C
Detalles adicionales : Weight : 0.10000kg
Descripción
This check key is compatible with any Hanna thermistor thermometer that uses the HI-765 series of probes. By simply connecting the calibration key to the probe input, the user can quickly determine if the meter needs to be recalibrated. If the reading on the display differs more than 0.4°C from the key rated value, the thermometer should be recalibrated.
- Designed to simulate a specific temperature set point
- For thermometers using HI-765 probes
- For periodic thermometer calibration, verify to at least two calibration points
- Check keys should be chosen based on the expected temperature of the sample
Especificaciones
Temperature probe calibration | |
0°C rated value | |
Thermistor thermometers with HI-765 Series Probes |
Calibration kep/plug | |
Yes | |
±0.4°C |