Description
- Measurement at specified temperature
- Including certificate
- Various points of measurement
Specifications
Specifications
| For Use With (Equipment) | BINDER Chambers Series MK |
| Description | Calibration certificate, temperature - temperature measurement incl. certificate and 27 measuring points (at specified temperature) |
| Calibration Points | 27 Measurement Points |
| Duration | 60 to 120 min. |
| Product Type | Calibration Certificate (Temperature) |
| For Use With (Application) | Temperature Measurement |