Learn More
BINDER™ Calibration Certificate for BINDER™ KB ECO Series
Shop All BINDER Products
Click to view available options
Calibration Type:
1 P Expanded Temperature
1 P Temperature
18 P Temperature
27 P Temperature
9 P Temperature
Unit Size:
Each
Description
- Calibration certificate
- For temperature
Specifications
Specifications
| For Use With (Equipment) | BINDER Chambers Series KB ECO |
| Description | Calibration certificate, temperature - temperature measurement incl. certificate, 15- 18 measuring points (at specified temperature) |
| Duration | 60 to 120 min. |
| Calibration Type | 18 P Temperature |
| Product Type | Calibration Certificate |