Learn More
Cadence Science™ Flexible Plastic (PTFE) Needles with Bite Protectors
Product Code. 17469373
Change view
Click to view available options
Gauge:
18G
Length (English):
1.5 in.
Diameter (Metric) Ball:
2 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gauge Length (English) Diameter (Metric) Ball Unit Size
17469373 18G 1.5 in. 2 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17469373 Supplier Cadence Science™ Supplier No. 9928B
Estimated Shipment: 18-02-2027
Covered at the hub end with 304 stainless steel tubing – protects tubing from damage due to biting during use.
Specifications
Disposable Single-use
For Use With (Application) Animal feeding
Gauge 18G
Sterility Sterile
Length (English) 1.5 in.
Diameter (Metric) Ball 2 mm