Learn More
Cadence Science™ Disposable Plastic (PTFE) Animal Feeding Needles
Product Code. 13244779
Change view
Click to view available options
Diameter (English) Ball:
0.074 in.
0.078 in.
Gauge:
18G
20G
Length (English):
1.2 in.
1.5 in.
2 in.
3 in.
Unit Size:
Pack of 100
Product Code. Diameter (English) Ball Gauge Length (English) unitSize
13244779 0.074 in. 20G 1.5 in. Pack of 100
17614825 0.078 in. 18G 3 in. Pack of 100
15786118 0.074 in. 20G 1.2 in. Pack of 100
15282035 0.078 in. 18G 2 in. Pack of 100
15282065 0.078 in. 18G 1.5 in. Pack of 100
5 options
Product Code. 13244779
Brand: Cadence Science™ 9920
For critical applications requiring sterile, flexible, single-use feeding needles
  • Nontoxic, nonpyrogenic
  • PTFE tubing is flexible and softer than other types of tubing
  • Low friction protects delicate organs
  • Translucent for easy confirmation of reagent delivery
  • Plastic luer hubs; soft silicone tips
  • Sterile, packaged in pouches
  • Five needles per pouch, 20 pouches per box
Specifications
Product Type Soft, Flexible Animal Feeding Needle
Material PTFE
Disposable Single-use
For Use With (Application) Animal feeding, For critical applications requiring sterile, flexible, single use feeding needles
Gauge 20G
Sterility Sterile
Length (English) 1.5 in.
Length (Metric) 0.38 cm
Diameter (English) Ball 0.074 in.
Diameter (Metric) Ball 0.019 cm