Learn More
C7 Mouse anti-Human, Unconjugated, Clone: 1H9C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66908-1-IG-150UL
This item is not returnable.
View return policy
Description
The C7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human C7. This antibody recognizes human antigen. The C7 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| C7 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 1H9C4 | |
| Unconjugated | |
| 150 μL | |
| 730 | |
| IgG1 |