missing translation for 'onlineSavingsMsg'
Learn More

C7 Mouse anti-Human, Unconjugated, Clone: 1H9C4, Proteintech

Product Code. p-7223157 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16849195 20 μL 20µL
16839195 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16849195

Brand: Proteintech 669081IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The C7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human C7. This antibody recognizes human antigen. The C7 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen C7
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1H9C4
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 730
Target Species Human
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.