Learn More
C7 Mouse anti-Human, Unconjugated, Clone: 1H9C4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66908-1-IG-20UL
 View more versions of this product
Product Code. 16849195
  • 1710.33 NOK / 20µL
Description
The C7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human C7. This antibody recognizes human antigen. The C7 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
C7
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1H9C4
Unconjugated
20 μL
730
IgG1