missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C-Reactive Protein/CRP Antibody (C6cc), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB200-442
This item is not returnable.
View return policy
Description
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunoassay, Sandwich ELISA, Immunoaffinity Purification.
Specifications
| C-Reactive Protein/CRP | |
| Monoclonal | |
| Unconjugated | |
| P02741 | |
| CRP | |
| Full length native protein (purified) (Human). | |
| 0.5 mg | |
| Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 1401 | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunoassay | |
| C6 | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoassay, Sandwich ELISA 1:100-1:2000 | |
| C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human CRP | |
| Store at 4C. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction