missing translation for 'onlineSavingsMsg'
Learn More

C-Reactive Protein/CRP Antibody (C6cc), Novus Biologicals™

Product Code. p-4974022 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.10mg
0.5mg
Product Code. Quantity unitSize
18259900 0.1 mg 0.10mg
18729163 0.5 mg 0.5mg
2 options
This item is not returnable. View return policy

Product Code. 18259900

Brand: Novus Biologicals NB2004420.1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunoassay, Sandwich ELISA, Immunoaffinity Purification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C-Reactive Protein/CRP
Applications ELISA, Immunohistochemistry, Immunoassay
Classification Monoclonal
Clone C6
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoassay, Sandwich ELISA 1:100-1:2000
Gene Accession No. P02741
Gene Alias C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895
Gene Symbols CRP
Host Species Mouse
Immunogen Full length native protein (purified) (Human).
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 1401
Test Specificity Human CRP
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.