Learn More
C-Reactive Protein/CRP Antibody (C6cc), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB200-442-0.1MG
This item is not returnable.
View return policy
Description
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunoassay, Sandwich ELISA, Immunoaffinity Purification.
Specifications
| C-Reactive Protein/CRP | |
| Monoclonal | |
| Unconjugated | |
| P02741 | |
| CRP | |
| Full length native protein (purified) (Human). | |
| 0.1 mg | |
| Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 1401 | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunoassay | |
| C6 | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoassay, Sandwich ELISA 1:100-1:2000 | |
| C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human CRP | |
| Store at 4C. Do not freeze. | |
| IgG2a |
Product Content Correction
Product Title
For Research Use Only