Learn More
BTX™ Electroporation Accessories: Banana Plug Connector
Description
- Compatible with most BTX Systems such as ECM 2001+ and Gemini
- See the electrode type for more details
Specifications
Specifications
| For Use With (Application) | Connect The Genetrode, Genepaddle, Microslide, Meander Fusion Chamber to The Electroporation Generator |
| For Use With (Equipment) | ECM 630, ECM 830, Gemini X2, Legacy ECM 2001 Electroporation Generators |
| Product Type | BTX Electroporation |
| Description | Micrograbber to Banana Plug Connection Cables, 10 ft |
| Length (English) Cable | 10 ft. |
| Length (Metric) Cable | 3.1 m |
| Sterility | Non-sterile |
| Propagating ccdB Vectors | Electroporation Accessory |
| Connection Type | Banana Plug to Micrograbber |
| Pulse Form | AC Sine Wave, DC Square Wave |