Learn More
Paragon Scientific™ BS/IP/SL(S) Viscometer for Transparent Liquids (UKAS ISO 17025 certified)
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
3016.67 NOK
Specifications
Calibrated | Yes |
---|---|
ASTM Numbers | D 445, D 446 |
Certifications/Compliance | ASTM D 445, D 446 and ISO 3104, 3105 |
Includes | UKAS ISO 17025 calibration |
Product Code | Brand | Viscosity Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Viscosity Range | Price | Quantity & Availability | |||||
16090380
|
Paragon Scientific™
SHORT01UKC |
1.05 min |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16010390
|
Paragon Scientific™
SHORT02UKC |
2.1 to 3 cSt |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030390
|
Paragon Scientific™
SHORT03UKC |
3.8 to 10 cSt |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050390
|
Paragon Scientific™
SHORT04UKC |
6 to 30 cSt |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070390
|
Paragon Scientific™
SHORT05UKC |
20 to 100 cSt |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16090390
|
Paragon Scientific™
SHORT06UKC |
60 to 300 cSt |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16010400
|
Paragon Scientific™
SHORT07UKC |
200 to 1000 cSt |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030400
|
Paragon Scientific™
SHORT08UKC |
600 to 3000 cSt |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050400
|
Paragon Scientific™
SHORT09UKC |
2000 to 10,000 cSt |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
- Supplied with full UKAS ISO 17025 calibration
Specifications
Yes | |
D 445, D 446 | |
ASTM D 445, D 446 and ISO 3104, 3105 | |
UKAS ISO 17025 calibration |
8 week lead-time