Learn More
MP Biomedicals™ Briggs N Salt Mixture, Powder
Product Code. p-7113097 Shop All MP Biomedicals Products
Click to view available options
Quantity:
2 kg
10 kg
50 kg
Unit Size:
10kg
2kg
50kg
Product Code. 11405632
Brand: MP Biomedicals™ 0290283402
Salt mixture for research animal diet.
  • Salt Mixture
  • For research animal diet
  • Powder
TRUSTED_SUSTAINABILITY
Specifications
Product Type Animal Diet
For Use With (Application) Research animal diet
Quantity 2 kg