missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD8 Mouse anti-Human, Clone: 5B6D6, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | BRD8 |
|---|---|
| Clone | 5B6D6 |
| Concentration | 1.47 mg/mL |
| Applications | Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16847085
|
Proteintech
60121-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16837085
|
Proteintech
60121-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Transcriptional repressor that contribute to growth arrest during terminal macrophage differentiation by repressing target genes involved in Ras-dependent proliferation. Represses MMP1 promoter activity.Specifications
| BRD8 | |
| 1.47 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| BRD8, bromodomain containing 8, p120, SMAP, SMAP2, TrCP120 | |
| BRD8 | |
| IgG1 | |
| Caprylic Acid Precipitation | |
| Antibody |
| 5B6D6 | |
| Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9H0E9 | |
| 10902 | |
| BRD8 Fusion Protein Ag0790 | |
| Primary | |
| -20°C | |
| BRD8 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title