missing translation for 'onlineSavingsMsg'
Learn More

BRD8 Mouse anti-Human, Clone: 5B6D6, Proteintech

Product Code. 16837085 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16837085 150 μL 150µL
16847085 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16837085

Brand: Proteintech 601211IG150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Transcriptional repressor that contribute to growth arrest during terminal macrophage differentiation by repressing target genes involved in Ras-dependent proliferation. Represses MMP1 promoter activity.

Specifications

Antigen BRD8
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5B6D6
Concentration 1.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene BRD8
Gene Accession No. Q9H0E9
Gene Alias BRD8, bromodomain containing 8, p120, SMAP, SMAP2, TrCP120
Gene Symbols BRD8
Host Species Mouse
Immunogen BRD8 Fusion Protein Ag0790
Purification Method Caprylic Acid Precipitation
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10902
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.