Learn More
Endecotts™ Brass Receiver
Use these receivers with test sieves. Endecotts™ Brass Receivers are widely used with test sieves for all types of laboratory sampling and particle size analysis
440.46 NOK
Description
- Brass construction
- For use with Brass Woven Wire Mesh Test Sieves
Specifications
Receiver | |
Brass | |
1 Ea. |