Learn More
Endecotts™ Brass Receiver
Use these receivers with test sieves. Endecotts™ Brass Receivers are widely used with test sieves for all types of laboratory sampling and particle size analysis
Brand:  Endecotts™ 100BRASREC
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12934265
Quantity Price
1 440.46 NOK / Each
10 404.56 NOK / Each
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Description
Description
  • Brass construction
  • For use with Brass Woven Wire Mesh Test Sieves
Specifications
Specifications
Receiver
100 mm
1 Ea.
Brass
For sieves