Learn More
Endecotts™ Brass Receiver
Use these receivers with test sieves. Endecotts™ Brass Receivers are widely used with test sieves for all types of laboratory sampling and particle size analysis
Brand: Endecotts™ 100BRASREC
Additional Details : Weight : 1.00000kg
Description
- Brass construction
- For use with Brass Woven Wire Mesh Test Sieves
Specifications
Receiver | |
100 mm | |
1 Ea. |
Brass | |
For sieves |