Learn More
Branson Ultrasonics™ Bransonic™ Ultrasonic Cleaning Bath Accessory, Support Racks
Description
For use with Branson ultrasonic baths
- Choice of sizes to fit all baths
Specifications
Specifications
| Type | Support Rack |
| For Use With (Equipment) | Ultrasonic Bath, Model 1800 |
| For Use With (Application) | For Model 1800 |
| Height (English) | 8 in. |
| Height (Metric) | 20.32 cm |
| Width (English) | 6 in. |