Learn More
Branson Ultrasonics™ Ultrasonic Cleaning Bath Accessory, Solid Insert Tray
Product Code. 13423639
Change view
Click to view available options
Capacity (Metric):
2.7 L
6.8 L
9.4 L
20.8 L
For Use With (Equipment):
Ultrasonic Bath, Model 1800
Ultrasonic Bath, Model 2800
Ultrasonic Bath, Model 3800
Ultrasonic Bath, Model 5800
Ultrasonic Bath, Model 8800
Unit Size:
Each
Product Code. Capacity (Metric) For Use With (Equipment) unitSize
13423639 6.8 L Ultrasonic Bath, Model 3800 Each
15141615 2.7 L Ultrasonic Bath, Model 1800 Each
13493629 2.7 L Ultrasonic Bath, Model 2800 Each
13473639 9.4 L Ultrasonic Bath, Model 5800 Each
13403649 20.8 L Ultrasonic Bath, Model 8800 Each
5 options
Product Code. 13423639
Brand: Branson Ultrasonics™ 100410174
Only null left
Add to basket
Add to basket
Solid tray for indirect cleaning method.
  • Solid tray for ultrasonic cleaners to prevent parts from touching tank bottom, also easily cleans discarded soil from solid tray
  • Also ideal for sample preparation
Specifications
Type Solid Insert Tray
Depth (English) 4 in.
Capacity (Metric) 6.8 L
Capacity (English) 1.5 gal.
Depth (Metric) 10.16 cm
For Use With (Equipment) Ultrasonic Bath, Model 3800
Includes Built in handle
Length (English) 8.5 in.
Length (Metric) 21.59 cm
Material Stainless Steel
Dimensions (L x W) 8.5 x 4.25 in. (21.59 x 10.79 cm)
Width (English) 4.25 in.
Width (Metric) 10.79 cm