Learn More
Branson Ultrasonics™ Bransonic™ Ultrasonic Cleaning Bath Accessory, Beakers
Product Code. 10041654
Change view
Click to view available options
Capacity (English):
8.4 oz.
20.28 oz.
Capacity (Metric):
250 mL
600 mL
Material:
Glass
Stainless Steel
Unit Size:
Each
Product Code. Capacity (English) Capacity (Metric) Material unitSize
10041654 20.28 oz. 600 mL Stainless Steel Each
15338644 8.4 oz. 250 mL Glass Each
15318644 20.28 oz. 600 mL Glass Each
3 options
Product Code. 10041654
Brand: Branson Ultrasonics™ 000410055
Only null left
Add to basket
Add to basket
For use with Bransonic ultrasonic baths
  • Suitable for all models
  • Available in glass, stainless steel, or polypropylene
Specifications
Type Beaker
Capacity (Metric) 600 mL
Capacity (English) 20.28 oz.
For Use With (Equipment) Ultrasonic Benchtop Cleaning Bath
Material Stainless Steel