Learn More
BRAND™ BRANDplates cellGrade™ 384-Well Flat-Bottom Microplates, PS
Product Code. 10407683 Shop All Brand Products
Change view
Click to view available options
Color:
White, Clear Bottom
Unit Size:
Pack of 50
Product Code. Color unitSize
10407683 White, Clear Bottom Pack of 50
1 options
Product Code. 10407683
Brand: BRAND™ 781986
Only null left
Add to basket
Add to basket
Sterile 384-well microplates with cellGrade™ surface treatment for optimal growth. Ideal for cultivating adherent cells with a hydrophilic PS surface featuring accessible carboxyl and hydroxyl groups. Enhances serum component binding for effective indirect cell adhesion.
  • Polystyrene construction
  • Sterile with included lids
  • Designed for the cultivation of adherent cell lines
  • Hydrophilic PS surface with accessible carboxyl and hydroxyl groups
  • Enhance serum component binding for indirect cell adhesion
  • Clearly distinguishable with orange color code and embossed alphanumeric coding for 384-well plates
  • BIO-CERT™ CELL CULTURE QUALITY: free from endotoxins, DNase, DNA, RNase, non-cytotoxic
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Certifications/Compliance ANSI, SLAS
Format Standard
Lid With Lid
Array 16 x 24
Autoclavable Not Autoclavable
Packaging 1/Pk., 50/Cs.
Binding Type Carboxyl and Hydroxyl Groups
Product Line BRANDplates
Color White, Clear Bottom
Product Type 384-well microplate
Material Polystyrene
Volume (Metric) Well 120 μL
No. of Wells 384
Sterility Sterile
Surface Treatment cellGrade
Total Volume (Metric) Well 120 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) Cultivation of adherent cell lines
No. per Case 50/Cs.
No. per Pack 1/Pk.
RNase Free Yes
Fit for Purpose Sterile