Learn More
BRAND™ BRANDplates cellGrade™ plus, 96-well Flat Bottom Microplates, PS
Product Code. 10564584 Shop All Brand Products
Change view
Click to view available options
Packaging:
1/Pk., 50/Cs.
5/Pk.
Color:
Black, Clear Bottom
Clear
White, Clear Bottom, Clear Bottom
Unit Size:
Pack of 5
Pack of 50
Product Code. Packaging Color unitSize
10564584 1/Pk., 50/Cs. White, Clear Bottom, Clear Bottom Pack of 50
15674017 5/Pk. White, Clear Bottom, Clear Bottom Pack of 5
10627713 - Clear Pack of 50
10793074 - Black, Clear Bottom Pack of 50
4 options
Product Code. 10564584
Brand: BRAND™ 782030
Only null left
Add to basket
Add to basket
Sterile 96-well microplates with cellGrade™plus surface treatment for optimal cell growth. Features protein-like surface for fast cell attachment and better yield. Ideal for cultivating sensitive or fastidious cell lines, even in reduced-serum media
  • Hydrophilic surface with freely-accessible amino, carboxyl and hydroxyl groups
  • Protein-like composition allows for direct cellular attachment
  • Sterilized by B-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • BIO-CERT™ CELL CULTURE QUALITY: free of pyrogens, endotoxins, human DNA, RNase, and DNase
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification
Order Info
Packs of 50 individually wrapped plates with lids
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Binding Property Hydrophilic
Certifications/Compliance ISO 11137, DIN EN ISO 10993
Format Standard
Lid With Lid
Array 8 x 12
Packaging 1/Pk., 50/Cs.
Color White, Clear Bottom, Clear Bottom
Product Type 96-well microplate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Sterile
Surface Treatment cellGrade Plus
Total Volume (Metric) Well 330 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) Cultivation of cells in reduced serum media and fastidious cell lines
No. per Case 50/Cs.
No. per Pack 1/Pk.
Well Color Clear