Learn More
Fisherbrand™ Borosilicate Glass Weighing Funnels
Product Code. 11572902 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
3 mL
6 mL
10 mL
Height (Metric):
70 mm
85 mm
100 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Height (Metric) Unit Size
11572902 3 mL 70 mm Each
11582902 6 mL 85 mm Each
11592902 10 mL 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11572902
In Stock
Add to basket
Edge
Add to basket
Introduce powder via the tubular stem. Fisherbrand™ Borosilicate Glass Weighing Funnels are ideal for weighing small quantities of powder.
Ideal for weighing small quantites of powder. Powder can be introduced via the tubular stem.
Specifications
Capacity (Metric) 3 mL
Material Borosilicate Glass
Height (Metric) 70 mm
For Use With (Application) Ideal for weighing small quantities of powder
Product Type Weighing Funnel