Learn More
Fisherbrand™ Glass Rectangular Coverslips
Shop All Fisherbrand Products
missing translation for 'orderingAttributeHoverText'
Thickness (Metric):
#1 (0.13 to 0.16 mm)
#1.5 (0.16 to 0.19 mm)
Length (Metric):
22 mm
24 mm
Width (Metric):
26 mm
32 mm
40 mm
50 mm
missing translation for 'unitSize'
Pack of 100
Pack of 200
Specifications
Specifications
| Product Type | Coverslip |
| Shape | Rectangle |
| Material | Soda Lime Glass |
| Length (Metric) | 22 mm |
| Width (Metric) | 26 mm |
| Thickness (Metric) | #1 (0.13 to 0.16 mm) |
| Quantity | 200/Pk. |