Specifications
| Product Type | Coverslip |
| Shape | Rectangle |
| Material | Soda Lime Glass |
| Length (Metric) | 22 mm |
| Width (Metric) | 40 mm |
| Thickness (Metric) | #1 (0.13 to 0.16 mm) |
| Quantity | 100 Pk. |