Specifications
| Product Type | Coverslip |
| Shape | Square |
| Material | Soda Lime Glass |
| Length (Metric) | 22 mm |
| Width (Metric) | 22 mm |
| Thickness (Metric) | #1.5 (0.16 to 0.19 mm) |
| Quantity | 200/Pk. |