Learn More
Fisherbrand™ Borosilicate Glass Narrow Neck Erlenmeyer Flasks
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
250 mL
300 mL
500 mL
1000 mL
2000 mL
3000 mL
5 L
Unit Size:
Each
Pack of 10
Specifications
Specifications
| Certifications/Compliance | ISO 1773 |
| Diameter (Metric) Outer Neck | 42 mm |
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass |
| Height (Metric) | 220 mm |
| Shape | Conical |
| Graduated | Yes |
| Mouth Style | Narrow |
| Diameter (Metric) Outer | 131 mm |