Learn More
Fisherbrand™ Borosilicate Glass Graduated Cylinder
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Graduations:
0.1 mL
0.2 mL
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
Unit Size:
Each
Pack of 2
Specifications
Specifications
| Material | Borosilicate Glass |
| Certifications/Compliance | DIN 12680, BS 604, ISO 4788 |
| Class | B |
| Capacity (Metric) | 100 mL |
| Scale | Blue Ceramic |
| Graduations | 1 mL |