Learn More
Velp Scientifica™ BOD Sensor
Product Code. 11795894 Shop All Velp Products
Change view
Click to view available options
Battery Type:
2 x 3 V Lithium
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Battery Type Unit Size
11795894 2 x 3 V Lithium Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11795894 Supplier Velp Scientifica™ Supplier No. F102B0133
  • ONLINE EXCLUSIVE PRICE
    2900.00 NOK
    2710.45 NOK / Pack of 1
    Save 189.55 NOK
    6% Off
Estimated Shipment: 02-12-2026
Velp Scientifica™ BOD Sensor is innovative, mercury-free and extremely reliable solution for BOD analysis (Biochemical Oxygen Demand).
Precise Control
  • Measurement on scales of 90, 250, 600 and 999 ppm BOD
  • Higher values can be measured by diluting the sample
  • Models with stirring stations offer VELP stirring quality
Extremely Practical
  • Fits directly on the bottle containing the sampl
  • Automatically stores 5 BOD values at 24-hour intervals
  • Enables analysis to continue over the weekend
  • Results directly readable at any time, even after five days
  • Direct readout in mg/l (ppm)
  • Compact stirring stations
Specifications
Battery Type 2 x 3 V Lithium
For Use With (Equipment) BOD Analysis