Learn More
Lovibond™ BOD Overflow Flask
For use with Lovibond oxygen measurment systems. Biochemical Oxygen Demand (BOD) measurement is used as a basis for the detection of biologically degradable organic matter in water.
Brand:  Lovibond™ 418664
Additional Details : Weight : 0.01500kg
 View more versions of this product
Product Code. 10529992
  • 354.00 NOK / Each
Estimated Shipment: 17-06-2024
Description
  • Overflow flask, 21.7 and 244mL
  • For accurate measurement of sample volumes
  • For BD 600, BD 606 and Oxi700 BOD Oxygen Meters
Specifications
Specifications
21.7 mL
BD 600, BD 606 and Oxi700 BOD Oxygen Meters
BOD Overflow Flask
BOD Analysis
0 to 400mg/L