Learn More
SI Analytics™ BlueLine pH Combination Electrode with Conical Membrane
Shop All SI Analytics Products
Click to view available options
Connector Types:
BNC
DIN
DIN, 4 mm Banana Plug
Metrohm Plug
Plug Head
Model:
Blueline 11 PH
Blueline 12 PH
Blueline 14 PH
Blueline 14 PH Id
Blueline 17 PH
Blueline 17 pH-R
Blueline 18 PH
Unit Size:
Each
Description
- Available in a variety of connection types
- Liquid electrolyte electrode
- For demanding measurements
- 120mm shaft length
- Type A glass conical membrane
Specifications
Specifications
| Connector Types | Plug Head |
| Junction | Platinum Diaphragm |
| Material | Glass |
| pH Range | 0 to 14 |
| Type | Combination pH Electrode |
| Internal Reference | Ag/AgCl |
| Membrane | Cone |
| Model | Blueline 11 PH |
| Temperature (Metric) Range | -5°C to +100°C |
| Length (Metric) | 120 mm |
| Show More |