Learn More
bis(4-Nitrophenyl) Sulfone, TRC
Product Code. 30240631
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240631 100 mg 100mg
30240910 50 mg 50mg
30240519 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30240631 Supplier Toronto Research Chemicals Supplier No. TRCB591915100MG
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1156-50-9
Molecular Formula C12H8N2O6S
Molecular Weight (g/mol) 308.26
Synonym Dapsone Impurity 11
IUPAC Name 1-nitro-4-(4-nitrophenyl)sulfonylbenzene
SMILES [O-][N+](=O)c1ccc(cc1)S(=O)(=O)c2ccc(cc2)[N+](=O)[O-]
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C12H8N2O6S/c15-13(16)9-1-5-11(6-2-9)21(19,20)12-7-3-10(4-8-12)14(17)18/h1-8H
Formula Weight 308.0103
Chemical Name or Material bis(4-Nitrophenyl) Sulfone