accessibility menu, dialog, popup

UserName

ONOSOONOO

CAS RN 1156-50-9

IUPAC Name: 1-nitro-4-(4-nitrobenzenesulfonyl)benzene
Synonyms: 1-nitro-4-(4-nitrobenzenesulfonyl)benzene
Molecular Weight (g/mol): 308.26
Molecular Formula: C12H8N2O6S
InChi Key: BVHNGWRPAFKGFP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)S(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results