missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-chlorophenylthio)methane, 99%, Thermo Scientific™

Product Code. 11469497
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469497 1 g 1g
11479497 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469497 Supplier Thermo Scientific Alfa Aesar Supplier No. B22018.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2393-97-7
Molecular Formula C13H10Cl2S2
Molecular Weight (g/mol) 301.24
MDL Number MFCD00041407
InChI Key NTZBITFHYUVXFR-UHFFFAOYSA-N
Synonym bis 4-chlorophenylthio methane, bis 4-chlorophenyl thio methane, 1-chloro-4-4-chlorophenyl sulfanylmethylsulfanyl benzene, 1-chloro-4-4-chlorophenyl sulfanyl methyl sulfanyl benzene, acmc-20alnn, bis-4-chlorophenylthio methane, methane, bis p-chlorophenyl-thio, bis-4-chlor-phenylmercapto-methan, 4-chloro-1-4-chlorophenylthio methylthio benzene
PubChem CID 137579
SMILES ClC1=CC=C(SCSC2=CC=C(Cl)C=C2)C=C1

Specifications

Melting Point 45°C to 48°C
Boiling Point 193°C to 195°C (2 mmHg)
Quantity 1 g
Beilstein 2216758
IUPAC Name 1-chloro-4-[(4-chlorophenyl)sulfanylmethylsulfanyl]benzene
Formula Weight 301.26
Percent Purity 99%
Chemical Name or Material Bis(4-chlorophenylthio)methane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.