missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-thiophenemethyl)amine hydrochloride, 99%, Thermo Scientific™

Product Code. 11415966
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415966 10 g 10g
11425966 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415966 Supplier Thermo Scientific Alfa Aesar Supplier No. B22290.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 336624-10-3
Molecular Formula C10H12ClNS2
Molecular Weight (g/mol) 245.783
MDL Number MFCD01812493
InChI Key RTJOHEYKSNEPMI-UHFFFAOYSA-N
Synonym bis 2-thiophenemethyl amine hydrochloride, bis thiophene-2-methyl amine hydrochloride, bis thiophen-2-ylmethyl amine hydrochloride, di-2-thenylamine hydrochloride, 1-thiophen-2-yl-n-thiophen-2-ylmethyl methanamine,hydrochloride, 1-thiophen-2-yl-n-thiophen-2-yl methyl methanamine-hydrogen chloride 1/1
PubChem CID 24212013
IUPAC Name 1-thiophen-2-yl-N-(thiophen-2-ylmethyl)methanamine;hydrochloride
SMILES C1=CSC(=C1)CNCC2=CC=CS2.Cl

Specifications

Melting Point ∼243°C (decomposition)
Quantity 10 g
Formula Weight 245.8
Percent Purity 99%
Chemical Name or Material Bis(2-thiophenemethyl)amine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.