accessibility menu, dialog, popup

UserName

SNHSHCl

CAS RN 336624-10-3

IUPAC Name: bis[(thiophen-2-yl)methyl]amine hydrochloride
Synonyms: bis(thiophen-2-ylmethyl)amine hydrochloride
Molecular Weight (g/mol): 245.78
Molecular Formula: C10H12ClNS2
InChi Key: RTJOHEYKSNEPMI-UHFFFAOYSA-N
SMILES: Cl.C(NCC1=CC=CS1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results