Learn More
ACROBiosystems Biotinylated Human BTLA (31-150) Protein, Fc, Avitag™
Biotinylated Protein
Brand: ACROBiosystems BTA-H82F3-25ug
Additional Details : Weight : 0.00010kg
Description
B- and T-lymphocyte attenuator (BTLA) is also known as B- and T-lymphocyte-associated protein, CD antigen CD272. BTLA contains one Ig-like V-type (immunoglobulin-like) domain. As a lymphocyte inhibitory receptor, BTLA / CD272 inhibits lymphocytes during immune response. BTLA / CD272 can interact with tyrosine phosphatases PTPN6/SHP-1 and PTPN11/SHP-2, and interact with TNFRSF14/HVEM.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 42.5 kDa. The protein migrates as 55-66 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
AAP44003.1 | |
25 μg | |
Lys 31 - Ser 150 | |
BTLA,CD272 | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
42.5 kDa | |
Biotinylated Human BTLA (31-150), Fc,Avitag (BTA-H82F3) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession # AAP44003.1). | |
-20°C | |
BTLA | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |