missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Biotinylated Human BTLA (31-150) Protein, Fc, Avitag™
Biotinylated Protein
5341.70 NOK - 23503.49 NOK
Specifications
Accession Number | AAP44003.1 |
---|---|
Molecular Weight (g/mol) | 42.5 kDa |
Source | Biotinylated Human BTLA (31-150), Fc,Avitag (BTA-H82F3) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession # AAP44003.1). |
Immunogen | Lys 31 - Ser 150 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16447239
|
ACROBiosystems
BTA-H82F3-200UG |
200 μg |
23503.49 NOK
200µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16457239
|
ACROBiosystems
BTA-H82F3-25UG |
25 μg |
5341.70 NOK
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
B- and T-lymphocyte attenuator (BTLA) is also known as B- and T-lymphocyte-associated protein, CD antigen CD272. BTLA contains one Ig-like V-type (immunoglobulin-like) domain. As a lymphocyte inhibitory receptor, BTLA / CD272 inhibits lymphocytes during immune response. BTLA / CD272 can interact with tyrosine phosphatases PTPN6/SHP-1 and PTPN11/SHP-2, and interact with TNFRSF14/HVEM.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 42.5 kDa. The protein migrates as 55-66 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
AAP44003.1 | |
Biotinylated Human BTLA (31-150), Fc,Avitag (BTA-H82F3) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession # AAP44003.1). | |
-20°C | |
BTLA | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
42.5 kDa | |
Lys 31 - Ser 150 | |
BTLA,CD272 | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |