Learn More
Thermo Scientific Biosafety Cabinet KI Test - Fisherbrand Biosafety Cabinets
Product Code. 30503418
Change view
Click to view available options
Product Type:
European KI Test
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30503418 European KI Test Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30503418 Supplier Thermo Scientific Supplier No. BSCKIFIS
 Restricted Item
Estimated Shipment: 29-12-2026
European KI Discus test for Fisherbrand biosafety cabinets to assess operator protection per standards and regulations.
  • Conducted in accordance with CEN BS EN12469:2000 (Table 5) for microbiological safety cabinets.
  • Evaluates operator/personal protection using the KI Discus test method.
  • Applicable during both commissioning and annual routine maintenance testing.
  • Aligned with national regulatory guidelines, including HSE (UK), which recommends annual testing.
In accordance with the European Standard for microbiological safety cabinets, CEN BS EN12469:2000 (Table - 5), the KI Discus test is stated as an optional test for operator/personal protection. This applies to both commissioning and annual routine maintenance testing. However, national regulations should always be adhered to as some national regulations such as those from the HSE (UK) recommend that the KI Discus test be performed at least annually.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Biosafety Cabinets
Duration 1 Year
Product Type European KI Test